siRNA Details
| Gene Symbol: Olfr1510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146431.2 | NP_666642.2 | 1 | 559 | ||
| GenBank mRNA | BC112967.1 | - | 502 | 40043789 | ||
| BC114920.2 | - | 502 | 40040851 | |||
| BC119377.1 | - | 1 | 559 | 8734126 | ||
| BC148301.2 | - | 502 | 40043795 | |||

Olfr1510
Genomic Map