siRNA Details
| Gene Symbol: Olfr1431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146414.1 | NP_666526.1 | 1 | 221 | ||
| GenBank mRNA | BC133710.1 | - | 1 | 225 | 40142241 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1431| Gene Symbol: Olfr1431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146414.1 | NP_666526.1 | 1 | 221 | ||
| GenBank mRNA | BC133710.1 | - | 1 | 225 | 40142241 | |