siRNA Details
| Gene Symbol: Olfr328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146502.2 | NP_666713.2 | 1 | 13 | ||
| GenBank mRNA | BC110465.2 | - | 1 | 13 | 40040838 | |
| BC110466.2 | - | 1 | 13 | 40040839 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr328| Gene Symbol: Olfr328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146502.2 | NP_666713.2 | 1 | 13 | ||
| GenBank mRNA | BC110465.2 | - | 1 | 13 | 40040838 | |
| BC110466.2 | - | 1 | 13 | 40040839 | ||