siRNA Details
| Gene Symbol: Olfr1462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146693.1 | NP_666904.1 | 1 | 911 | ||
| GenBank mRNA | BC128026.1 | - | 1 | 915 | 40136583 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1462| Gene Symbol: Olfr1462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146693.1 | NP_666904.1 | 1 | 911 | ||
| GenBank mRNA | BC128026.1 | - | 1 | 915 | 40136583 | |