siRNA Details
Gene Symbol: Olfr694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146452.2 | NP_666663.2 | 1 | 28 | ||
GenBank mRNA | BC129796.1 | - | 1 | 28 | 40042951 | |
BC129797.1 | - | 1 | 28 | 40042952 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr694 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146452.2 | NP_666663.2 | 1 | 28 | ||
GenBank mRNA | BC129796.1 | - | 1 | 28 | 40042951 | |
BC129797.1 | - | 1 | 28 | 40042952 |