siRNA Details
| Gene Symbol: Olfr463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146413.2 | NP_666525.2 | 1 | 294 | ||
| GenBank mRNA | BC106969.1 | - | 1 | 294 | 40041195 | |
| BC106970.1 | - | 1 | 292 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr463| Gene Symbol: Olfr463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146413.2 | NP_666525.2 | 1 | 294 | ||
| GenBank mRNA | BC106969.1 | - | 1 | 294 | 40041195 | |
| BC106970.1 | - | 1 | 292 | |||