siRNA Details
| Gene Symbol: Vmn1r189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145844.1 | NP_665843.1 | 1 | 869 | ||
| GenBank mRNA | BC150715.1 | - | 1 | 968 | 9087626 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r189| Gene Symbol: Vmn1r189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145844.1 | NP_665843.1 | 1 | 869 | ||
| GenBank mRNA | BC150715.1 | - | 1 | 968 | 9087626 | |