siRNA Details
Gene Symbol: Olfr934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146442.1 | NP_666653.1 | 1 | 583 | ||
GenBank mRNA | BC113124.1 | - | 1 | 582 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146442.1 | NP_666653.1 | 1 | 583 | ||
GenBank mRNA | BC113124.1 | - | 1 | 582 |