siRNA Details
| Gene Symbol: Olfr345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146945.1 | NP_667156.1 | 1 | 548 | ||
| GenBank mRNA | BC148219.1 | - | 1 | 550 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr345| Gene Symbol: Olfr345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146945.1 | NP_667156.1 | 1 | 548 | ||
| GenBank mRNA | BC148219.1 | - | 1 | 550 | ||