siRNA Details
| Gene Symbol: Olfr376 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172686.1 | NP_001166157.1 | 3 | 270 | ||
| NM_146922.2 | NP_667133.1 | 2 | 139 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr376| Gene Symbol: Olfr376 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172686.1 | NP_001166157.1 | 3 | 270 | ||
| NM_146922.2 | NP_667133.1 | 2 | 139 | |||