siRNA Details
Gene Symbol: Olfr376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172686.1 | NP_001166157.1 | 3 | 339 | ||
NM_146922.2 | NP_667133.1 | 2 | 208 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172686.1 | NP_001166157.1 | 3 | 339 | ||
NM_146922.2 | NP_667133.1 | 2 | 208 |