siRNA Details
| Gene Symbol: Olfr20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146923.2 | NP_667134.2 | 2 | 1019 | ||
| GenBank mRNA | BC138172.1 | - | 1 | 922 | 8861191 | |
| BC138173.1 | - | 1 | 922 | 8861192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr20| Gene Symbol: Olfr20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146923.2 | NP_667134.2 | 2 | 1019 | ||
| GenBank mRNA | BC138172.1 | - | 1 | 922 | 8861191 | |
| BC138173.1 | - | 1 | 922 | 8861192 | ||