siRNA Details
| Gene Symbol: E430018J23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198011.2 | NP_932128.2 | 3 | 935 | ||
| GenBank mRNA | AK088474.1 | - | 3 | 899 | ||
| BC010442.1 | - | 933 | 3586722 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E430018J23Rik| Gene Symbol: E430018J23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198011.2 | NP_932128.2 | 3 | 935 | ||
| GenBank mRNA | AK088474.1 | - | 3 | 899 | ||
| BC010442.1 | - | 933 | 3586722 | |||