siRNA Details
| Gene Symbol: 1700029I15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183112.3 | NP_898935.1 | 1, 2 | 142 | ||
| GenBank mRNA | AK006499.1 | - | 1, 2 | 143 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700029I15Rik| Gene Symbol: 1700029I15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183112.3 | NP_898935.1 | 1, 2 | 142 | ||
| GenBank mRNA | AK006499.1 | - | 1, 2 | 143 | ||