siRNA Details
| Gene Symbol: Gsdmc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183194.3 | NP_899017.2 | 8, 9 | 942 | ||
| GenBank mRNA | AK037079.1 | - | 8, 9 | 943 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC099685.1 | - | 986 | Chr. - - [-] on Build | ||

Gsdmc3
Genomic Map