siRNA Details
| Gene Symbol: Hamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183257.3 | NP_899080.1 | 3 | 280 | ||
| GenBank mRNA | AK007975.1 | - | 3 | 270 | ||
| AK008993.1 | - | 3 | 281 | |||
| AY232841.1 | - | 3 | 269 | |||
| BC086895.1 | - | 3 | 277 | 6765293 | ||
| D17962.1 | - | 1 | 85 | |||

Hamp2
Genomic Map