siRNA Details
| Gene Symbol: AF067061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199060.1 | NP_951015.1 | 608 | |||
| GenBank mRNA | AF067061.1 | - | 607 | |||
| AF067062.1 | - | 608 | ||||
| BC115951.1 | - | 1 | 1054 | 40049464 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AF067061| Gene Symbol: AF067061 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199060.1 | NP_951015.1 | 608 | |||
| GenBank mRNA | AF067061.1 | - | 607 | |||
| AF067062.1 | - | 608 | ||||
| BC115951.1 | - | 1 | 1054 | 40049464 | ||