siRNA Details
| Gene Symbol: Fam178b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201365.2 | NP_958753.2 | 10 | 1536 | ||
| GenBank mRNA | AK141223.1 | - | 10 | 1492 | ||
| BC050210.1 | - | 10 | 1439 | 30009484 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam178b| Gene Symbol: Fam178b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201365.2 | NP_958753.2 | 10 | 1536 | ||
| GenBank mRNA | AK141223.1 | - | 10 | 1492 | ||
| BC050210.1 | - | 10 | 1439 | 30009484 | ||