siRNA Details
| Gene Symbol: Cyp2c54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206537.2 | NP_996260.1 | 1 | 78 | ||
| GenBank mRNA | AI181879.1 | - | 1 | 67 | ||
| BF659738.1 | - | 1 | 78 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2c54| Gene Symbol: Cyp2c54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206537.2 | NP_996260.1 | 1 | 78 | ||
| GenBank mRNA | AI181879.1 | - | 1 | 67 | ||
| BF659738.1 | - | 1 | 78 | |||