siRNA Details
| Gene Symbol: AB124611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206536.2 | NP_996259.2 | 2, 3 | 460 | ||
| GenBank mRNA | AB124612.1 | - | 2, 3 | 360 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AB124611| Gene Symbol: AB124611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206536.2 | NP_996259.2 | 2, 3 | 460 | ||
| GenBank mRNA | AB124612.1 | - | 2, 3 | 360 | ||