siRNA Details
| Gene Symbol: Vmn1r168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166842.1 | NP_001160314.1 | 1 | 508 | ||
| Gene Symbol: Vmn1r177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206872.2 | NP_996755.2 | 1 | 508 | ||

Vmn1r168
Genomic Map