siRNA Details
| Gene Symbol: NIT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020202.4 | NP_064587.1 | 7 | 611 | ||
| GenBank mRNA | AF260334.1 | - | 7 | 610 | ||
| AF284574.1 | - | 7 | 558 | |||
| AK225979.1 | - | 6 | 1089 | |||
| AK295835.1 | - | 6 | 478 | |||
| AK313704.1 | - | 7 | 553 | |||
| BC020620.1 | - | 7 | 568 | 4775062 | ||
| BC107890.1 | - | 7 | 599 | 6648910 | ||
| FJ224326.1 | - | 7 | 558 | |||

NIT2
Genomic Map