siRNA Details
| Gene Symbol: Olfr325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207153.2 | NP_997036.2 | 1 | 954 | ||
| GenBank mRNA | BC141015.1 | - | 1 | 978 | 9055627 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr325| Gene Symbol: Olfr325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207153.2 | NP_997036.2 | 1 | 954 | ||
| GenBank mRNA | BC141015.1 | - | 1 | 978 | 9055627 | |