siRNA Details
| Gene Symbol: Olfr320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207230.1 | NP_997113.1 | 1 | 754 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr320| Gene Symbol: Olfr320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207230.1 | NP_997113.1 | 1 | 754 | ||