siRNA Details
| Gene Symbol: Olfr358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207235.1 | NP_997118.1 | 1 | 120 | ||
| GenBank mRNA | BC119215.1 | - | 1 | 155 | 8733964 | |
| BC119217.1 | - | 1 | 155 | 8733966 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr358| Gene Symbol: Olfr358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207235.1 | NP_997118.1 | 1 | 120 | ||
| GenBank mRNA | BC119215.1 | - | 1 | 155 | 8733964 | |
| BC119217.1 | - | 1 | 155 | 8733966 | ||