siRNA Details
| Gene Symbol: Olfr684 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207249.1 | NP_997132.1 | 1 | 23 | ||
| GenBank mRNA | BC120636.1 | - | 1 | 62 | 40129559 | |
| BC120638.1 | - | 1 | 62 | 40129561 | ||
More Information
Set Membership: |
|

Olfr684
Genomic Map