siRNA Details
| Gene Symbol: Kcnk18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207261.3 | NP_997144.1 | 1 | 316 | ||
| GenBank mRNA | AK139600.1 | - | 1 | 750 | ||
| AK162136.1 | - | 1 | 319 | |||
| AY325301.1 | - | 1 | 230 | |||
| AY542902.1 | - | 1 | 230 | |||
| BC104132.1 | - | 1 | 230 | |||
| BC104133.1 | - | 1 | 227 | |||
| BC115705.1 | - | 1 | 229 | |||
| BC115887.1 | - | 1 | 223 | |||
| BC127136.1 | - | 1 | 230 | 40045157 | ||
| BC127137.1 | - | 1 | 230 | 40045158 | ||

Kcnk18
Genomic Map