siRNA Details
| Gene Symbol: Vmn1r174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207548.2 | NP_997431.2 | 1 | 250 | ||
| Gene Symbol: Vmn1r173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166718.1 | NP_001160190.1 | 1 | 250 | ||

Vmn1r174
Genomic Map