siRNA Details
| Gene Symbol: Olfr199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207550.2 | NP_997433.2 | 1 | 286 | ||
| GenBank mRNA | BC128028.1 | - | 1 | 290 | 40136725 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr199| Gene Symbol: Olfr199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207550.2 | NP_997433.2 | 1 | 286 | ||
| GenBank mRNA | BC128028.1 | - | 1 | 290 | 40136725 | |