siRNA Details
Gene Symbol: Olfr372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207555.2 | NP_997438.1 | 1 | 939 | ||
GenBank mRNA | BC150671.1 | - | 1 | 939 | 9087582 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207555.2 | NP_997438.1 | 1 | 939 | ||
GenBank mRNA | BC150671.1 | - | 1 | 939 | 9087582 |