siRNA Details
| Gene Symbol: Olfr1040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207561.2 | NP_997444.2 | 1 | 893 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1040| Gene Symbol: Olfr1040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207561.2 | NP_997444.2 | 1 | 893 | ||