siRNA Details
| Gene Symbol: Olfr1173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207566.1 | NP_997449.1 | 1 | 649 | ||
| GenBank mRNA | BC104085.2 | - | 1 | 649 | 40042097 | |
| BC104278.1 | - | 1 | 649 | 40041579 | ||
| BC104279.1 | - | 1 | 649 | 40041581 | ||
| BC127020.1 | - | 1 | 649 | 40042733 | ||
| BC134355.1 | - | 1 | 649 | 40041582 | ||

Olfr1173
Genomic Map