siRNA Details
| Gene Symbol: Hist3h2bb-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206882.1 | NP_996765.1 | 1 | 35 | ||
| GenBank mRNA | BC107301.1 | - | 1 | 34 | 40053183 | |
| BC107302.1 | - | 1 | 35 | 40053184 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist3h2bb-ps| Gene Symbol: Hist3h2bb-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_206882.1 | NP_996765.1 | 1 | 35 | ||
| GenBank mRNA | BC107301.1 | - | 1 | 34 | 40053183 | |
| BC107302.1 | - | 1 | 35 | 40053184 | ||