siRNA Details
| Gene Symbol: Klra17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133203.5 | NP_573466.3 | 4 | 483 | ||
| GenBank mRNA | AB033769.1 | - | 4 | 496 | ||
| AB190787.1 | - | 3 | 342 | |||
| AK080021.1 | - | 4 | 484 | |||
| BC132236.1 | - | 4 | 444 | 40130513 | ||

Klra17
Genomic Map