siRNA Details
| Gene Symbol: Gm9758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198666.3 | NP_941068.1 | 5 | 917 | ||
| GenBank mRNA | BC048454.1 | - | 5 | 917 | 6741646 | |
| Gene Symbol: Speer4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122661.1 | NP_001116133.1 | 5 | 926 | ||
| GenBank mRNA | AY151402.1 | - | 209 | |||
| BC100449.1 | - | 872 | ||||

Gm9758
Genomic Map