siRNA Details
| Gene Symbol: Olfr151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207664.2 | NP_997547.1 | 1 | 557 | ||
| GenBank mRNA | BC138117.1 | - | 1 | 557 | 8861136 | |
| BC138118.1 | - | 1 | 557 | 8861137 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr151| Gene Symbol: Olfr151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207664.2 | NP_997547.1 | 1 | 557 | ||
| GenBank mRNA | BC138117.1 | - | 1 | 557 | 8861136 | |
| BC138118.1 | - | 1 | 557 | 8861137 | ||