siRNA Details
| Gene Symbol: Zfp729a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183146.3 | NP_898969.2 | 4 | 388 | ||
| GenBank mRNA | AK040978.1 | - | 4 | 426 | ||
| AK138962.1 | - | 4 | 388 | |||
| BC098180.1 | - | 4 | 400 | 5401052 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp729a| Gene Symbol: Zfp729a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183146.3 | NP_898969.2 | 4 | 388 | ||
| GenBank mRNA | AK040978.1 | - | 4 | 426 | ||
| AK138962.1 | - | 4 | 388 | |||
| BC098180.1 | - | 4 | 400 | 5401052 | ||