siRNA Details
| Gene Symbol: Olfr118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213721.2 | NP_998886.2 | 1 | 35 | ||
| GenBank mRNA | BC128012.1 | - | 1 | 39 | 40136025 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr118| Gene Symbol: Olfr118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_213721.2 | NP_998886.2 | 1 | 35 | ||
| GenBank mRNA | BC128012.1 | - | 1 | 39 | 40136025 | |