siRNA Details
Gene Symbol: BC117090 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001332.2 | NP_001001332.2 | 3 | 261 | ||
GenBank mRNA | AY169235.1 | - | 3 | 261 | ||
BC117064.1 | - | 3 | 261 | 40126383 | ||
BC117090.1 | - | 3 | 261 | 40126409 |
Gene Symbol: BC100530 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082546.1 | NP_001076015.1 | 3 | 294 | ||
GenBank mRNA | BC100530.1 | - | 3 | 472 | 30289147 |
Gene Symbol: Stfa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082543.1 | NP_001076012.1 | 3 | 273 | ||
GenBank mRNA | BY113926.1 | - | 3 | 273 | ||
M92417.1 | - | 3 | 242 |
