siRNA Details
| Gene Symbol: BC117090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001332.2 | NP_001001332.2 | 1 | 57 | ||
| GenBank mRNA | AY169235.1 | - | 1 | 57 | ||
| BC117064.1 | - | 1 | 57 | 40126383 | ||
| BC117090.1 | - | 1 | 57 | 40126409 | ||

BC117090
Genomic Map