siRNA Details
| Gene Symbol: Olfr145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146313.1 | NP_666425.1 | 1 | 922 | ||
| GenBank mRNA | BC137773.1 | - | 1 | 1003 | 8860792 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr145| Gene Symbol: Olfr145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146313.1 | NP_666425.1 | 1 | 922 | ||
| GenBank mRNA | BC137773.1 | - | 1 | 1003 | 8860792 | |