siRNA Details
| Gene Symbol: Olfr872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146560.2 | NP_666771.2 | 2 | 1011 | ||
| GenBank mRNA | BC120630.1 | - | 1 | 929 | 40129553 | |
| BC120632.1 | - | 1 | 929 | 40129555 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr872| Gene Symbol: Olfr872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146560.2 | NP_666771.2 | 2 | 1011 | ||
| GenBank mRNA | BC120630.1 | - | 1 | 929 | 40129553 | |
| BC120632.1 | - | 1 | 929 | 40129555 | ||