siRNA Details
| Gene Symbol: Gm13023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007077.2 | NP_001007078.2 | 4 | 1528 | ||
| GenBank mRNA | AK135972.1 | - | 4 | 1511 | ||
| BC084594.1 | - | 4 | 1509 | 30478700 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13023| Gene Symbol: Gm13023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007077.2 | NP_001007078.2 | 4 | 1528 | ||
| GenBank mRNA | AK135972.1 | - | 4 | 1511 | ||
| BC084594.1 | - | 4 | 1509 | 30478700 | ||