siRNA Details
| Gene Symbol: AY761184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007582.3 | NP_001007583.1 | 2 | 326 | ||
| GenBank mRNA | AK008498.1 | - | 2 | 332 | ||
| AY761184.1 | - | 2 | 281 | |||
| EU760895.1 | - | 1 | 122 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AY761184| Gene Symbol: AY761184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007582.3 | NP_001007583.1 | 2 | 326 | ||
| GenBank mRNA | AK008498.1 | - | 2 | 332 | ||
| AY761184.1 | - | 2 | 281 | |||
| EU760895.1 | - | 1 | 122 | |||