siRNA Details
| Gene Symbol: Wdr91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013366.1 | NP_001013384.1 | 12 | 1726 | ||
| GenBank mRNA | BC043682.1 | - | 7 | 995 | ||
| BC055046.1 | - | 12 | 1726 | 6488548 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr91| Gene Symbol: Wdr91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013366.1 | NP_001013384.1 | 12 | 1726 | ||
| GenBank mRNA | BC043682.1 | - | 7 | 995 | ||
| BC055046.1 | - | 12 | 1726 | 6488548 | ||