siRNA Details
| Gene Symbol: Olfr23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010970.1 | NP_035100.1 | 1 | 618 | ||
| GenBank mRNA | X89688.1 | - | 1 | 244 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr23| Gene Symbol: Olfr23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010970.1 | NP_035100.1 | 1 | 618 | ||
| GenBank mRNA | X89688.1 | - | 1 | 244 | ||