siRNA Details
| Gene Symbol: Tmprss13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013373.2 | NP_001013391.2 | 13 | 2164 | ||
| GenBank mRNA | BC010843.1 | - | 5 | 1017 | ||
| BC042878.1 | - | 12 | 1904 | |||
| BC085323.1 | - | 13 | 2153 | 30734093 | ||

Tmprss13
Genomic Map