siRNA Details
| Gene Symbol: Vmn2r123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009485.1 | NP_033511.1 | 6 | 1853 | ||
| GenBank mRNA | AF011411.1 | - | 6 | 1853 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r123| Gene Symbol: Vmn2r123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009485.1 | NP_033511.1 | 6 | 1853 | ||
| GenBank mRNA | AF011411.1 | - | 6 | 1853 | ||