siRNA Details
| Gene Symbol: Nrsn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009948.1 | NP_001009948.1 | 3 | 908 | ||
| GenBank mRNA | AK140190.1 | - | 3 | 925 | ||
| BC088982.1 | - | 3 | 940 | 5700110 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrsn2| Gene Symbol: Nrsn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009948.1 | NP_001009948.1 | 3 | 908 | ||
| GenBank mRNA | AK140190.1 | - | 3 | 925 | ||
| BC088982.1 | - | 3 | 940 | 5700110 | ||