siRNA Details
| Gene Symbol: Gm128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024841.3 | NP_001020012.1 | 1 | 963 | ||
| GenBank mRNA | AB690575.1 | - | 1 | 835 | ||
| AK144488.1 | - | 1 | 964 | |||
| BC092540.1 | - | 1 | 1060 | 6742207 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm128| Gene Symbol: Gm128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024841.3 | NP_001020012.1 | 1 | 963 | ||
| GenBank mRNA | AB690575.1 | - | 1 | 835 | ||
| AK144488.1 | - | 1 | 964 | |||
| BC092540.1 | - | 1 | 1060 | 6742207 | ||